ENA024295495 Enamine

1-[2-(4-chlorophenyl)ethyl]-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD06356468
  • Cas Number : 790232-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09459

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09459
Availability details for
Quantity
  • Availability
  • Add to Cart