ENA024295498 Enamine

1-benzyl-3-(pyridin-3-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06356503
  • Cas Number : 956959-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09462
Availability details for
Quantity
  • Availability
  • Add to Cart