ENA024295500 Enamine

2-(2-chloroethanesulfonyl)-1,4-dimethylbenzene

  • MDL number MFCD06356455
  • Cas Number : 790232-11-0
  • Molecular Weight : 232.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09464
Availability details for
Quantity
  • Availability
  • Add to Cart