ENA024295505 Enamine

2-chloro-1-(2,5-dimethoxyphenyl)ethan-1-one

  • MDL number MFCD03701176
  • Cas Number : 1204-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-77
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09469
Availability details for
Quantity
  • Availability
  • Add to Cart