ENA024295508 Enamine

5-(5-ethyl-4,5,6,7-tetrahydro-1-benzothien-2-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06356495
  • Cas Number : 790232-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09472
Availability details for
Quantity
  • Availability
  • Add to Cart