ENA024295509 Enamine

5-(5-tert-butyl-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06660722
  • Cas Number : 885460-60-6
  • Molecular Weight : 294.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09473
Availability details for
Quantity
  • Availability
  • Add to Cart