ENA024295510 Enamine

5-(5-tert-butyl-4,5,6,7-tetrahydro-1-benzothien-2-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06356497
  • Cas Number : 790232-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09474

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09474
Availability details for
Quantity
  • Availability
  • Add to Cart