ENA024295535 Enamine

2-chloro-N-{3-[(pyrrolidin-2-ylidene)sulfamoyl]phenyl}acetamide

  • Cas Number : 790270-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-153
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09499
Availability details for
Quantity
  • Availability
  • Add to Cart