ENA024295536 Enamine

2-[(4-benzyl-5-methyl-1H-imidazol-2-yl)sulfanyl]acetic acid

  • MDL number MFCD06358024
  • Cas Number : 790272-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-163
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09500
Availability details for
Quantity
  • Availability
  • Add to Cart