ENA024295541 Enamine

2-{8-oxatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5-tetraen-4-yloxy}acetic acid

  • MDL number MFCD03066368
  • Cas Number : 34855-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09505
Availability details for
Quantity
  • Availability
  • Add to Cart