ENA024295549 Enamine

2-[4-(3-chlorobenzenesulfonyl)piperazine-1-carbonyl]benzoic acid

  • MDL number MFCD05263278
  • Cas Number : 743445-14-9
  • Molecular Weight : 408.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09513
Availability details for
Quantity
  • Availability
  • Add to Cart