ENA024295562 Enamine

2-methoxybenzenecarbothioamide

  • MDL number MFCD04973324
  • Cas Number : 42590-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09526

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-09526
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart