ENA024295579 Enamine

2-cyclohexaneamido-4H,5H,6H-cyclopenta[b]thiophene-3-carboxylic acid

  • MDL number MFCD06357357
  • Cas Number : 790263-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09543
Availability details for
Quantity
  • Availability
  • Add to Cart