ENA024295580 Enamine

2-[(2,2-dimethylpropanoyl)amino]-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid

  • MDL number MFCD06357376
  • Cas Number : 790263-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09544
Availability details for
Quantity
  • Availability
  • Add to Cart