ENA024295591 Enamine

3-chloro-4-(difluoromethoxy)-5-methoxybenzaldehyde

  • MDL number MFCD05270919
  • Cas Number : 747411-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09555
Availability details for
Quantity
  • Availability
  • Add to Cart