ENA024295595 Enamine

1-sec-butyldihydropyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06357377
  • Cas Number : 651718-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09559
Availability details for
Quantity
  • Availability
  • Add to Cart