ENA024295610 Enamine

2,4-dichloro-5-[(pentan-2-yl)sulfamoyl]benzoic acid

  • MDL number MFCD05263644
  • Cas Number : 743451-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09574
Availability details for
Quantity
  • Availability
  • Add to Cart