ENA024295613 Enamine

3-[3-(4-chlorophenyl)-4-formyl-1H-pyrazol-1-yl]propanenitrile

  • MDL number MFCD02705847
  • Cas Number : 255710-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09577

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09577
Availability details for
Quantity
  • Availability
  • Add to Cart