ENA024295631 Enamine

1-{4-methoxy-3-[(pyrrolidin-1-yl)methyl]phenyl}ethan-1-one

  • MDL number MFCD06357354
  • Cas Number : 790263-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09595
Availability details for
Quantity
  • Availability
  • Add to Cart