ENA024295646 Enamine

2-chloro-N-(2-fluoro-4-methylphenyl)acetamide

  • MDL number MFCD06356481
  • Cas Number : 366-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09610
Availability details for
Quantity
  • Availability
  • Add to Cart