ENA024295678 Enamine

5-benzyl-4-propyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD05134119
  • Cas Number : 663181-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09642
Availability details for
Quantity
  • Availability
  • Add to Cart