ENA024295685 Enamine

2-{[2-(propan-2-yl)phenoxy]methyl}oxirane

  • MDL number MFCD06357351
  • Cas Number : 5904-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09649
Availability details for
Quantity
  • Availability
  • Add to Cart