ENA024295688 Enamine

2-{[(4-methylphenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD02755455
  • Cas Number : 58511-20-9
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09652
Availability details for
Quantity
  • Availability
  • Add to Cart