ENA024295692 Enamine

4-[(phenylsulfanyl)methyl]benzoic acid

  • MDL number MFCD03991405
  • Cas Number : 88382-49-4
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09656
Availability details for
Quantity
  • Availability
  • Add to Cart