ENA024295707 Enamine

4-{4-[2-chloro-5-(trifluoromethyl)benzenesulfonyl]piperazin-1-yl}-4-oxobut-2-enoic acid

  • Cas Number : 743445-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09671
Availability details for
Quantity
  • Availability
  • Add to Cart