ENA024295715 Enamine

4-[(chloroacetyl)amino]-N,N-diethylbenzamide

  • MDL number MFCD05273650
  • Cas Number : 746608-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09679

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09679
Availability details for
Quantity
  • Availability
  • Add to Cart