ENA024295724 Enamine

2-chloro-N-{[(2,6-dimethylphenyl)carbamoyl]methyl}acetamide

  • MDL number MFCD05273566
  • Cas Number : 50333-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09688
Availability details for
Quantity
  • Availability
  • Add to Cart