ENA024295725 Enamine

3-(2-methoxyphenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06357392
  • Cas Number : 956625-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09689
Availability details for
Quantity
  • Availability
  • Add to Cart