ENA024295748 Enamine

3-[2,5-dimethyl-1-(2,2,2-trifluoroethyl)-1H-pyrrol-3-yl]-3-oxopropanenitrile

  • MDL number MFCD06335113
  • Cas Number : 750604-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09712
Availability details for
Quantity
  • Availability
  • Add to Cart