ENA024295760 Enamine

2-(4-chlorophenyl)-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD06335185
  • Cas Number : 21278-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09724

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09724
Availability details for
Quantity
  • Availability
  • Add to Cart