ENA024295763 Enamine

2-{[4-(4-ethoxyphenyl)-5-(2-methoxyphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD06335177
  • Cas Number : 750607-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09727
Availability details for
Quantity
  • Availability
  • Add to Cart