ENA024295770 Enamine

2-amino-N-cyclopropylbenzamide

  • MDL number MFCD03015446
  • Cas Number : 30510-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09734

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09734
Availability details for
Quantity
  • Availability
  • Add to Cart