ENA024295792 Enamine

N-(2-fluorophenyl)-2-mercaptoacetamide

  • MDL number MFCD00046833
  • Cas Number : 217317-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09756
Availability details for
Quantity
  • Availability
  • Add to Cart