ENA024295796 Enamine

4-oxo-4-(3-phenyl-4,5-dihydro-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD06357383
  • Cas Number : 790263-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09760
Availability details for
Quantity
  • Availability
  • Add to Cart