ENA024295810 Enamine

5-bromo-2-methoxybenzene-1-carbothioamide

  • MDL number MFCD06335754
  • Cas Number : 303111-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09774
Availability details for
Quantity
  • Availability
  • Add to Cart