ENA024295813 Enamine

N-(2,3-dimethylcyclohexyl)urea

  • MDL number MFCD06335205
  • Cas Number : 953807-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09777
Availability details for
Quantity
  • Availability
  • Add to Cart