ENA024295814 Enamine

(2-methylcyclohexyl)urea

  • MDL number MFCD06335204
  • Cas Number : 750608-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-09778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09778
Availability details for
Quantity
  • Availability
  • Add to Cart