ENA024295819 Enamine

2-(2-chlorobenzamido)-4,5-dimethoxybenzoic acid

  • MDL number MFCD02031957
  • Cas Number : 402609-67-0
  • Molecular Weight : 335.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09783
Availability details for
Quantity
  • Availability
  • Add to Cart