ENA024295824 Enamine

1-(5-chloro-1-benzofuran-2-yl)ethan-1-one

  • MDL number MFCD00661909
  • Cas Number : 1646-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09788
Availability details for
Quantity
  • Availability
  • Add to Cart