ENA024295830 Enamine

3-(1H-indol-3-yl)-5-(4-methoxyphenyl)-4,5-dihydro-1H-pyrazol-4-ol

  • MDL number MFCD04638810
  • Cas Number : 736949-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09794
Availability details for
Quantity
  • Availability
  • Add to Cart