ENA024295841 Enamine

2-ethoxy-2,2-diphenylacetic acid

  • MDL number MFCD00426098
  • Cas Number : 7495-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-116
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09805
Availability details for
Quantity
  • Availability
  • Add to Cart