ENA024295848 Enamine

2-methyl-4-(4-methylpiperazin-1-yl)aniline

  • MDL number MFCD06335528
  • Cas Number : 16154-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09812
Availability details for
Quantity
  • Availability
  • Add to Cart