ENA024295860 Enamine

{[4-(chlorosulfonyl)benzoyl]amino}acetic acid

  • MDL number MFCD06340099
  • Cas Number : 155493-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09824
Availability details for
Quantity
  • Availability
  • Add to Cart