ENA024295877 Enamine

4-butoxy-3-chloro-5-ethoxybenzoic acid

  • MDL number MFCD06337009
  • Cas Number : 749920-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09841
Availability details for
Quantity
  • Availability
  • Add to Cart