ENA024295885 Enamine

3-(2,4-dimethylphenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06357834
  • Cas Number : 956386-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09849
Availability details for
Quantity
  • Availability
  • Add to Cart