ENA024295886 Enamine

(3-phenyl-1,2,4-oxadiazol-5-yl)acetonitrile

  • MDL number MFCD06357815
  • Cas Number : 57459-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27...29
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09850

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09850
Availability details for
Quantity
  • Availability
  • Add to Cart