ENA024295887 Enamine

2-[4-(chloromethyl)-1,3-thiazol-2-yl]pyridine

  • MDL number MFCD06336996
  • Cas Number : 342025-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09851
Availability details for
Quantity
  • Availability
  • Add to Cart