ENA024295898 Enamine

3-(2,5-dimethoxyphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06357439
  • Cas Number : 956206-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09862

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09862
Availability details for
Quantity
  • Availability
  • Add to Cart