ENA024295899 Enamine

3-(2,4-dimethylphenyl)-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06357442
  • Cas Number : 956744-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-09863
Availability details for
Quantity
  • Availability
  • Add to Cart