ENA024295920 Enamine

2-[(4-ethoxyphenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD03119353
  • Cas Number : 4394-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09884
Availability details for
Quantity
  • Availability
  • Add to Cart